Alexander Raspatory, OR Grade, Sklar®
Supplier: Sklar
55-5980
10197-602EA 594.08 USD
10197-602
Alexander Raspatory, OR Grade, Sklar®
Raspatories
Sklar's® Alexander Raspatory, also known as the Alexander-Farabeuf Pediatric Raspatory, is commonly used for scraping periosteum, cartilage, and tissue from bone.
  • Latex free and reusable
It is a double-ended instrument with one chisel end and one double mallet shaped end. The mallet shaped end is actually has two gouge-style tips that are also used for periosteum removal.