Alexander Pediatric Raspatory, OR Grade, Sklar®
Supplier: Sklar
22-7560
10195-218EA 567.88 USD
10195-218
Alexander Pediatric Raspatory, OR Grade, Sklar®
Raspatories
Sklar's® Alexander Pediatric Raspatory, also known as the Alexander-Farabeuf Pediatric Raspatory, is commonly used for scraping periosteum, cartilage, and tissue from bone.
  • Reusable
  • Latex-free
It is a double-ended instrument with one chisel end and one double mallet shaped end
The mallet shaped end is actually has two gouge-style tips that are also used for periosteum removal. This product is straight with a 10×30 mm tip and a length of 6".