Econo™ Olsen-Hegar Needle Holder, Floor Grade, Sklar
Supplier: Sklar
21-877
10195-060BX 166.98 USD
10195-060
Econo™ Olsen-Hegar Needle Holder, Floor Grade, Sklar
Forceps
The Econo™ Olsen-Hegar Needle Holder is a ratcheted, finger ring instrument that also has built-in scissors.
The Econo™ Olsen-Hegar Needle Holder is used with intermediate weight sutures, commonly in plastic surgery procedures. Econo™ instruments are floor grade, disposable instruments sold in bulk quantities.