Would you like to log in again?
Silicone Heat Transfer Fluid 710, FoxWest Sales
Supplier: FOXWEST PC
2301
10611-550
Silicone Heat Transfer Fluid 710, FoxWest Sales
Silicone Heat Transfer Fluid 710 is a Dimethyl-Diphenylsiloxane silicone fluid designed for use as a high temperature bath fluid or high temperature heat transfer fluid
The working temperature range for silicone bath fluid is 25 to 250°C (77 to 482°F) in an open system, and 25 to 300°C (77 to 572°F) in a closed system. The fluid viscosity is 400cSt.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty