Would you like to log in again?
DNase Displace, FoxWest Sales
Supplier: FOXWEST PC
1409
10611-536
DNase Displace, FoxWest Sales
DNase Displace removes DNase and unwanted DNA
It can be used on all surfaces, including glass, plastic, metal, and ceramic. Phosphate-free, safely disposable, non-toxic, and totally rinsable for the decontaminated surface.
It works instantly, eliminating the time-consuming method of autoclaving.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty