BCR® Full Finger Polyester Glove Liners, Large, Berkshire
Supplier: Berkshire
BGL2.8LB
89233-406CS 1569.17 USD
89233-406
BCR® Full Finger Polyester Glove Liners, Large, Berkshire
Gloves
BCR full finger polyester glove liners, large (blue cuff).
  • Full line offers a choice to better provide even the most discriminating wearer with the most appropriate glove liner
  • Due to specific engineering, BCR® Glove Liners fit most hands up to a size 9
  • This allows for easier stock control, while the availability of a larger size accommodates the hard to fit hands
Composition and attributes: 100% continuous filament, knitted polyester. Composition allows moisture to be absorbed and wicked away from skin during extended wear. Constructed using special seamless techniques. No-run and non-ravel. Ambidextrous for easy stock control. Launderable for multiple reuse. The glove liner is suitable in any ISO class cleanroom when worn under a nitrile, latex, vinyl, neoprene or other type glove.
BCR® Polyester Glove Liners - Protection and comfort during extended wear. BCR® Glove Liners are the choice in any industry, when the job calls for long hours of use when hand comfort is a concern. Special fiber construction allows for moisture absorption during extended wear.
Packaging: 10 per pack, 20 packs/case. Quality control and lot traceability identification on each case. Packaging is recyclable.