Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Glass Inserts, MicroSolv

Supplier: MicroSolv Technology Corp
9504S-04ND 9504S-01ND 9504S-02ND 9504S-03ND 9502S-02L-K 95096-707-RS 95096-7-RSAQL 9504S-04N 9504S-03N 95096-707 9504S-02N 9504S-01N 9502S-05ND




 

ORDER

Expand All / Collapse All

Glass
Description Size Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Inserts for 9mm Screw or 11mm Snap/Crimp Top (2mL) 12 x 32mm Autosampler Vials 6 x 31 mm 200 µL 9502S-02L-K 97052-762 Case of 1,000 Retrieving
Inserts for 96 Well Plates 8 x 40 mm 700 µL 95096-707 97055-518 Pack of 250 Retrieving

Borosilicate Glass
Description Size Details Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Inserts for 9504S Vials 6 x 38 mm Small Volume, Conical 250 µL 9504S-02N 97055-348 Pack of 100 Retrieving
Inserts for 15 x 45mm (4mL) Vials 6 x 38 mm Conical with Attached Springs 300 µL 9504S-04N 97055-356 Pack of 100 Retrieving
Inserts for 9504S Vials 8 x 40 mm Small Volume, High Precision with Plastic Springs 700 µL 9504S-03N 97055-352 Pack of 100 Retrieving

RSA Borosilicate Glass
Description Size Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Inserts for 96 Well Plates 8 x 40 mm 700 µL 95096-707-RS 10804-760 Pack of 250 Retrieving
Inserts for 96 Well Plates in Quick Load Tray 8 x 40 mm 700 µL 95096-7-RSAQL 10804-762 Pack of 96 Retrieving

Type 33 Borosilicate Glass
Description Size Details Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Inserts for 15 x 45mm (4mL) Vials 8 x 40 mm Precision with Mandrel Tip 250 µL 9504S-01N 97055-344 Pack of 100 Retrieving

Deactivated Silanized Glass
Description Size Details Capacity Supplier No. VWR Catalog Number Unit Price Quantity
Inserts for 15 x 45mm (4mL) Vials 6 x 38 mm Precision with Mandrel Tip 250 µL 9504S-01ND 97055-346 Pack of 100 Retrieving
Inserts for 9504S Vials 6 x 38 mm Small Volume, High Precision 250 µL 9504S-02ND 97055-350 Pack of 100 Retrieving
Inserts for 15 x 45mm (4mL) Vials 6 x 38 mm Conical with Attached Springs 300 µL 9504S-04ND 97055-358 Pack of 100 Retrieving
Inserts for 9mm (2mL) Screw or Snap/Crimp Top Autosampler Vials 6 x 31 mm Flat Bottom 350 µL 9502S-05ND 97052-458 Pack of 100 Retrieving
Inserts for 9504S Vials 8 x 40 mm Small Volume, High Precision with Plastic Springs 700 µL 9504S-03ND 97055-354 Pack of 100 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty