Would you like to log in again?
Rubber Pipette Fillers, Argos Technologies
Supplier: COLE-PARMER INSTRUMENT COMPANY
4395-03 4395-02 4395-01 P6005BK
10834-150EA 58.93 USD InStock
10834-150 10834-142 10834-138 10834-136
Rubber Pipette Fillers, Argos Technologies
Pipets Pipet Filler Bulbs
Manufactured for the safe and reliable aspiration and delivery of laboratory liquids
Bulbs are produced from thicker rubber than competitive offerings in order to give users a more robust feel, and provide greater pipetting control.
Recommended for use with volatile or dangerous liquids, where contamination needs to be avoided.
Pipetting bulbs are manufactured from natural rubber and are ideal for use with serological pipettes up to 50mL. The pinch and release valve system allows for one-hand operation. Each valve has a chemically-resistant, stainless steel ball bearing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty