Would you like to log in again?
Hoses, Huber
Supplier: Huber
9613 9619 9617 9614 9616 9618 6488 9615
10221-926 10222-096 10221-410 10221-420 10221-416 10221-422 10221-786 10222-094
Hoses, Huber
PTFE hose with braided metal reinforcement.
Special insulation with additional smooth silicon hose, ends covered with silicon caps.
Nominal width: 20 or 25 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty