Would you like to log in again?
Pressure Booster, Huber
Supplier: Huber
527.0016 527.0014 527.0018 527.0017
10221-138EA 7269.54 USD InStock
10221-138 10221-132 10221-134 10221-136
Pressure Booster, Huber
Pumps Compressors
External pressure pump.
  • All parts in the liquid circuit and housing made of stainless steel
  • Operation either manual (on/off) or external via 3–line–connector
Bearing out of the liquid circuit. Cable (Option) for the potentional free contact of Unistat. Tango and the big power Unistats, all Compatible Control Thermostats and Unichillers.
Temperature range: –120 to 300 °C
Electrical: 460 V, 3~ 60 Hz
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty