Would you like to log in again?
LTP125TOC Cleanroom Sampling Swab, Berkshire
Supplier: BERKSHIRE CORP
LTP125TOC.25
89496-570CS 901.25 USD InStock
89496-570
LTP125TOC Cleanroom Sampling Swab, Berkshire
Swabs Swabs for Sampling
The LTP125TOC large, knitted polyester swab has been specifically designed for Total Organic Carbon analysis and TOC tests in low-TOC environments.
  • Low particles, fibers, ions, and extractables
  • Good chemical resistance
  • Soft and non-abrasive with no contaminating adhesives
  • Cleanroom Class 100 to 100000
It offers a grooved handle for easy snapping. Each QC lot is certified with ultra-low TOC levels <50 ppb. Autoclavable. Free from silicone, amides, and DOP. Contamination control, with critically low particles, fibers, ions, and extractables.
Applications include total organic carbon analysis and TOC test procedure applications for validation in low-TOC environments. TOC sampling surfaces for cleaning validation in many industries including medical device, in vitro diagnostics, pharmaceutical, reusable medical devices.
Ordering information: 21 swabs per pack, 25 packs per case (double pack).
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty