Would you like to log in again?
MagBind® EquiPure NGS Library and gDNA Normalization Kits, Omega Bio-tek
Supplier: OMEGA BIO-TEK
M6423-00 M6423-01 M6445-00 M6423-00 M6445-00 M6423-01 M6445-01 M6445-01
CA10820-632 10820-634 10820-636 10820-632 CA10820-636 CA10820-634 10820-638 CA10820-638
MagBind® EquiPure NGS Library and gDNA Normalization Kits, Omega Bio-tek
Protein Linking and Labeling Systems
  • DNA output concentrations +/- 10%
  • No quantification required
  • Normalizes genomic DNA or NGS libraries
Many high-throughput applications, such as sequencing, including Next Generation Sequencing (NGS) and genotyping, require the input DNA concentration to be within a certain range for optimal results. Traditionally, a tedious process of quantification, calculation, and concentration adjustment must be carried out to normalize the DNA samples.
The MagBind EquiPure Normalizer Kits completely eliminate the need to quantify and aliquot DNA, saving time, quantification bias and tip costs. Using proprietary MagBind Normalizer Beads and binding buffer system, input DNA of various quantities is simply bound, washed and eluted to a final normalized product. The magnetic beads have a limited binding capacity and therefore allow a predefined amount of DNA to be captured and eluted.
The MagBind EquiPure Normalizer Kits are fully automatable on multiple liquid handling platforms including Hamilton STAR, Tecan Evo, Caliper Sciclone, and Beckman Coulter Biomek instruments. Additionally, these kits allow users to adjust the quantity of DNA that binds to the magnetic beads and the elution volumes can be adjusted to vary DNA concentrations, providing flexibility for multiple needs.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty