Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Pre-wetted Sealed Edge Cleanroom Laundered 100% Polyester Knit Wiper, Berkshire

Supplier: BERKSHIRE CORP
SPXHA.001B.8
SatPax® HA-R combines ValuSeal®-HA ultrasonically sealed edge cleanroom laundered wipers composed of 100% continuous filament polyester knit fabric wetted with 70% IPA/30% DIW

A high solution saturation level of 65% makes SatPax® HA-R ideal for regulated applications or applications requiring higher solution levels delivered to the surface. Laundered and packaged in ISO Class 4 cleanroom. Re-sealable solvent resistant packaging.

Increases cleaning protocol consistency. Flatpack packaging reduces packaging waste by 28% compared to j-folded packaging. Designed for use in wet cleaning of critical surfaces where control of flammable solvents and flammable solvent concentrations is required.

This pre-wetted format provides a cost effective and easy to use solution versus traditional bulk handling of solvents, maintenance of squirt bottles and inconsistent wetting and cleaning associated and inconsistent wetting and cleaning associated with wetting a dry wiper during cleaning

Certifications: Complies with RoHS and REACH requirements.

Ordering information: 25 Sheets per pack,82 packs per case. Double packed to ensure cleanliness. Quality control and lot traceability identification on each case.Lot Number embossed on each pack. Packaging is recyclable.




 

ORDER

Expand All / Collapse All
W x L Packaging Supplier No. VWR Catalog Number Unit Price Quantity
30.5 x 30.5 cm (12 x 12") C-Folded SPXHA.01B.8 89215-524 Case of 1,200 Retrieving
Small Business Enterprise
View MSDS SDS
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty