Would you like to log in again?
Gamma Wipe® MicroSeal®-VP CapSure VP Sterile Sealed Edge Cleanroom Laundered Wiper, Berkshire
Supplier: BERKSHIRE CORP
GWCPSVP.ST25
89136-878
Gamma Wipe® MicroSeal®-VP CapSure VP Sterile Sealed Edge Cleanroom Laundered Wiper, Berkshire
Gamma Wipe® MicroSeal®-VP is a sterile sealed-edge, cleanroom-laundered 100% polyester knit wiper for ISO class 3 and above controlled environments
Laundered and packaged in ISO class 4 cleanroom. Ultrasonically sealed edge for reduced fiber contamination. Gamma irradiated and sterile validated to a 10-6 SAL per AAMI guidelines.
Chemically compatible with common cleaning and disinfecting solutions, including isopropyl alcohol, phenols, bleaches and quarternary ammonium compounds. Designed for cleaning sterile surfaces and equipment during aseptic processing. Excellent for cleaning sterile laminar air flow units, barrier isolators and bio-containment units.
Each package is gamma irradiated sterile to a 10-6 Sterility Assurance Level. The Gamma Wipe® MicroSeal®-VP offers critically low particle, fiber, ion, and extracble levels as well as abrasion resistance and absorbency. It is chemically compatible with common cleaning and disinfecting solutions, including including isopropyl alcohol, phenols, bleaches, and quarternary ammonium compounds
Certifications: ANSI/AAMI/ISO procedures have been followed for verification of irradiation dose with quarterly audits being performed to monitor bioburden levels and dose. Complies with RoHS 2002/05 and REACH requirements. Meets cGMP requirements for traceability.
Ordering information: 20 sheets per pack, 25 packs per case (double packed). Quality control and lot traceability identification on each case. Certificate of sterility, expiration date and lot number on each pack.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty