Would you like to log in again?
Calibration Bend, Huber
Supplier: Huber
9779 9914 9925 9369 324776
10221-658 10221-636 10221-664 10221-660 10221-662
Calibration Bend, Huber
Calibration bend mounted on the machine outlet
The calibration bend has a sensor pocket for sensor which has to be calibrated by the user. The measured values appears on the display as reference for the internal flow temperature sensor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty