Would you like to log in again?
Bath, Huber
Supplier: Huber
6056 6058 6055 6059 6215 6057 6060 6329
10221-278 10221-282 10221-276 10221-284 10222-090 10221-280 10221-286 10222-092
Bath, Huber
Insulated stainless steel baths are available in 3 standard sizes and can be customised to suit requirements
These insulated stainless steel baths can be fitted with inlet and outlet connections directly into the bath (direct temperature control) or into the double–wall for indirect temperature control are available.
The drain is on the short side as standard but can be fitted on the long side on request. The order number has the suffix L (e.g. 6052–L).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty