Would you like to log in again?
Axygen® Reagent Resevoir, Axygen® Scientific
Supplier: CORNING AXYGEN
RESMW8LPSI RES-SW8-HP-S RES-MW8-LP-SI RESSW384FXSI
Axygen® offers a complete line of single well and multiple well reagent reservoirs which are available in numerous formats for automated and manual applications
All reservoirs are designed to comply with ANSI/SLAS 1-2004: Microplates - Footprint Dimensions, and are compatible with most automation systems. Manufactured with virgin polypropylene for excellent chemical resistance.
ORDER
Expand All / Collapse All
Description Capacity Color Sterility Packaging Supplier No. VWR Catalog Number Unit Price Quantity
Multi-Well Reagent Reservoir with 8 Channel Trough, Low Profile 290 mL Clear Sterile Individually Wrapped RES-MW8-LP-SI 95042-900 Case of 25 Retrieving
Single Well Reagent Reservoir with 384 Bottom Troughs, Medium Profile 170 mL Clear Sterile Individually Wrapped RESSW384FXSI 89179-660 Case of 25 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty