Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

VWR® Series Solid Door Stainless Steel Laboratory Freezers with Natural Refrigerant

Supplier: VWR International
VWR®
AFSP-23 AFSP-49
The VWR® Series Solid Door Stainless Steel Laboratory Freezers come standard with microprocessor temperature controllers allowing for the precise temperature management necessary for critical cold storage.

  • Stainless steel construction
  • All new temperature display and alarm module
  • Door ajar and power failure alarms
  • Up-to-the-minute air and sample simulated temperature information
  • Swivel casters (adds 5" to height; included in dimensions)

The all new temperature display and alarm module offers a comprehensive array of features, providing up-to-the-minute air and sample simulated temperature information, as well as the added notification features of the door ajar and power failure alarms.

Remote alarm contacts enable end users to tie these units into existing alarm systems to notify appropriate personnel of alarm events as soon as possible.

Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.

Certifications: UL/C-UL listed.

Ordering information: Two-year parts and labor warranty, five years on compressor parts.



 

ORDER

Expand All / Collapse All
Electrical Volume Exterior Dimensions Defrost Door Type No. of Shelves Shipping Weight VWR Catalog Number Unit Price Quantity
115V, 60Hz, 1HP, 9.6A 1388 L (49 cu. ft.) 54W x 31½D x 78¼"H Auto 2 Swing Stainless 6 Adjustable 515 lbs. 10819-632 Each Retrieving
Product and/or packaging is safer to human and environmental health
Product is energy and/or water efficient Product available on GSA Advantage® UL Listed cUL Listed
115V, 60Hz, ½HP, 3.7A 651 L (23 cu. ft.) 27W x 313/8D x 78¼"H Auto 1 Swing Stainless 3 Adjustable 310 lbs. 10820-418 Each Retrieving
Product and/or packaging is safer to human and environmental health
Product is energy and/or water efficient Product available on GSA Advantage® UL Listed cUL Listed
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Environmentally Preferable Attribute Product and/or packaging is safer to human and environmental health, Product is energy and/or water efficient

ACCESSORIES

Expand All / Collapse All

Accessories
Description VWR Catalog Number Unit Price Quantity
VWR® Additional Full Shelf for 1388 L (49 cu. ft.) Units 10819-636 Each Retrieving
VWR® Additional Full Shelf for 651 L (23 cu. ft.) Units 10819-634 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

COMPLEMENTARY PRODUCTS

Expand All / Collapse All

Complementary Products
Description Range Accuracy Fill Solution VWR Catalog Number Unit Price Quantity
Thermometer with One Bottle Probe -58.00 to 158.00°F (-50.00 to 70.00°C) ±0.30°C Glycol 89094-746 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SERVICES


Services: Extended One- or Three-Year Warranty options are available for VWR®Solid Door Stainless Steel Laboratory Freezers. Must be purchased at the same time as the equipment. All service plans are valid only at the point of equipment sale.

White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer’s receiving dock.
Expand All / Collapse All

Service Options
Description VWR Catalog Number Unit Price Quantity
VWR® Extended One-Year Warranty (Only Valid at Point of Unit Sale) VWR® Solid Door Stainless Steel Laboratory Freezers 10819-642 Each Retrieving
VWR® Extended Three-Year Warranty (Only Valid at Point of Unit Sale) for VWR® Solid Door Stainless Steel Laboratory Freezers 10819-644 Each Retrieving

Services
Description Details Includes Service Coverage Out-of-Service Coverage VWR Catalog Number Unit Price Quantity
VWR® NIST On-Site Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. Calibration at a single point using NIST traceable standards. Upon completion, a calibration form will be provided with “as found” and “as left” measurements. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote. 75875-880 Each Retrieving
VWR® NIST On-Site PM and Calibration Each device has been powered up and set temperature point for 24 hours before on-site service. PM and Calibration service includes cleaning of seals, fans, and gaskets. The chamber is inspected interiorly and exteriorly. After PM, technicians will calibrate the chamber as described in Calibration section. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote. 75875-888 Each Retrieving
VWR® NIST Traceable Validate (IOQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. Validation (IQ/OQ) includes a standardized protocol procedure, calibration, tuning, IQ (verify design spec, system SOP, environmental conditions etc), and OQ (Perform empty chamber temperature mapping, open door and power loss testing). (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote. 75875-950 Each Retrieving
VWR® NIST Traceable Validate (IOPQ) IQ does not include installation. Unit must be powered up and set temperature point for 24 hours before on-site service. IQ/OQ will be performed as described in the (IQ/OQ) section. The PQ consists of loaded chamber temperature mapping for an additional 24 hours. (50 Miles From Center City) Philadelphia, PA; New York, NY; Boston, MA; Washington, DC; Raleigh, NC; Tampa, FL; Miami, FL; Atlanta, GA; Nashville, TN; Cleveland, OH; Chicago, IL; Indianapolis, IN; Minneapolis, MN; Milwaukee, WI; St. Louis, MO; Dallas, TX; Houston, TX; Austin, TX; Denver, CO; Salt Lake City, UT; Phoenix, AZ; San Diego, CA; Los Angeles, CA; Las Vegas, NV; Portland, OR; Seattle, WA. Other service/area/brand, please visit www.vwr.com/equip_services or call 888.793.2300 to get a quote. 75875-942 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty