Would you like to log in again?
VWR® Performance Series Solid Door Stainless Steel Auto Defrost Laboratory Freezers with Natural Refrigerant
Supplier: VWR International
VWR®
AFPP-49-TS AFPP-23-TS
10819-494EA 17965.81 USD InStock
10819-494 10819-492
VWR® Performance Series Solid Door Stainless Steel Auto Defrost Laboratory Freezers with Natural Refrigerant
Freezers Laboratory/General Purpose Freezers
The VWR® Performance Series Solid Door Stainless Steel Auto Defrost Laboratory Freezers feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies.
  • Data acquisition
  • 24/7 data logging
  • 24-hour digital temp chart
  • Min/max temp history
  • USB port for data transfer
  • 4-20mA output
  • Touchscreen technology
  • Digital temperature display
  • Thermometer bar
  • Electronic owner’s manual access
  • Troubleshooting guide
  • Password protected
  • Swivel casters (adds 5" to height; included in dimensions)
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings
Standard Features Package. Keyed Door Locks, Casters, Sample-Simulated Probe, Probe Access Port, UL/C-UL Listed
Multi-point Temperature Mapping via NIST traceable data acquisition, 15 probes in air and 3 probes in sample solution for 360 minutes with temperature readings every 60 seconds.
Environmentally friendly, natural hydrocarbon (HC) refrigerants (not containing hydrofluorocarbons) vastly reduce global-warming potential (GWP) while meeting new EPA/SNAP mandates and UL, ASHRAE compliance guidelines.
An automatic cycle-defrost design prevents product damaging temperature spikes.
Certifications: UL/C-UL listed.
Ordering information: Units are supplied iwth a three-year parts and labor warranty, plus an additional two-year compressor parts warranty.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
COMPLEMENTARY PRODUCTS
SERVICES
Services: Extended One- or Three-Year Warranty options are available for VWR® Performance Solid Door Stainless Steel Auto Defrost Laboratory Freezers. Must be purchased at the same time as the equipment. All service plans are valid only at the point of equipment sale.
White Glove Installation and Verification Service–the white glove installation includes unpacking, inspection, removal/disposal of packing materials, transporting order via elevator and merchandise set up in the room of your choice, installation of casters or leveling feet, power and electrical connections inspected, supply voltage verified, functional and performance verification documented. Our engineers will move the unit from the customer's receiving dock to the lab within the same building. This service does NOT include transportation from vendor warehouse to customer’s receiving dock.
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty