Mycotic Nail Nipper, OR Grade, Sklar
Supplier: Sklar
97-1057
10193-952EA 173.57 USD
10193-952
Mycotic Nail Nipper, OR Grade, Sklar
Cutters
Sklar's mycotic nail nipper is a heavy, podiatry instrument used for cutting thickened, mycotic nails.
  • Double-action mechanism
  • Allows the hardened nail to be trimmed more easily with less effort
  • Reusable and latex-free
  • Premium OR-grade
  • Non sterile
The barrel spring and concave blades allow for the jaws to open wide, enabling a more accurate placement of the sharp blades. This device also features a locking mechanism at the bottom of the handle to keep the blades locked in place when they are not in use.