Would you like to log in again?
Solar Scope Education Version
Supplier: SOLARSCOPE LLC SE
04P102
470008-022EA 319.95 USD InStock
470008-022
Solar Scope Education Version
Telescopes
The user-friendly Solarscope lets users safely view the light of the sun.
  • Students Calculate Earth's Orbital Characteristics
  • Guidebook Included
  • Length: 25½”
  • Width: 18”
  • Height: 15”
Students can observe solar sun spots, planet transits, and eclipses of the moon with this instrument. Using the Solarscope, students can also calculate earth's rotation speed, earth's orbit ellipticity, the sun’s rotation speed, and more.
The telescope is easy to align and manipulate, and it includes a user’s guide and teacher’s workbook.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty