Would you like to log in again?
Jackson Original Laryngectomy Tube, Disposable Grade, Sklar
Supplier: Sklar
96-2815 96-2816 96-2813 96-2814 96-2817 96-2818
Sklar's Jackson Original Style Laryngectomy Tube is similar to the Jackson original style tracheostomy tube, yet shorter in overall length
The original style tube has a different locking mechanism than the improved style, and primarily impacts removal of the inner cannula for cleaning. The tube style that is utilized can best be determined by the physician, patient, and caregiver.
A laryngectomy tube consists of one obturator, an inner cannula, and an outer cannula, but is intended to provide a more permanent airway than a tracheostomy tube. It is commonly placed following the removal of the patient's larynx, often due to severe trauma, such as cancer.
ORDER
Expand All / Collapse All
Description Size Material Supplier No. VWR Catalog Number Unit Price Quantity
Jackson Original Laryngectomy Tube #4 Stainless Steel 96-2813 10160-096 Case of 25 Retrieving Restricted
Jackson Original Laryngectomy Tube #5 Stainless Steel 96-2814 10160-074 Case of 12 Retrieving Restricted
Jackson Original Laryngectomy Tube #6 Stainless Steel 96-2815 10160-076 Case of 12 Retrieving Restricted
Jackson Original Laryngectomy Tube #7 Stainless Steel 96-2816 10160-078 Case of 12 Retrieving Restricted
Jackson Original Laryngectomy Tube #8 Stainless Steel 96-2817 10160-080 Case of 12 Retrieving Restricted
Jackson Original Laryngectomy Tube #9 Stainless Steel 96-2818 10160-082 Case of 12 Retrieving Restricted
restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:
  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address
* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty