Would you like to log in again?
TC Adson Dressing Forceps, OR Grade, Sklar
Supplier: Sklar
51-3447 51-3447C
89025-098EA 262.77 USD InStock
89025-098 75847-650
TC Adson Dressing Forceps, OR Grade, Sklar
Forceps
Tungsten Carbide Adson Dressing Forceps are thumb forceps used for holding dressing materials such as cotton and gauze during surgical procedures, changing dressings, or packing wounds.
  • Wide thumb grasp
  • Gold handles
TC instruments are designed to stay sharp for a significantly longer period of time than other standard instruments
Censitrac Ready products are industry standard and constructed of stainless steel.
Sklar permanently bonds tungsten carbide inserts to the instrument, which maximizes control and precision.
Ordering information: Censitrac instruments are custom barcoded to reflect the facility. This is unique customization. The initial order for the facility can take 14 to 30 days, subsequent orders can take less time.
Caution: Customers must have a Censitrac Site ID to purchase Censitrac instruments.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty