Would you like to log in again?
Deep Well Plates, Axygen® Scientific
Supplier: Corning
P-96-450V-C P-96-450-R-C P-96-450-V-C P-LID-PS P-96-450-VCS P-ADAPTER-1 P-96-450R-C P-96-450R-C P-1ML-SQ-C-BC P-96-450V-CS
CA47743-996 47743-982 47743-996 80086-086 47743-998 89080-512 490005-358 CA47743-982 490013-264 CA47743-998
Deep Well Plates, Axygen® Scientific
Microplates HTS Microplates
Axygen® Deep Well and Assay plates are used for sample collection, in vitro growth chambers, and long-term storage.
Made of polypropylene, they offer excellent chemical resistance and will withstand temperatures down to –80 °C.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty