Human Recombinant BCA-1 (from E. coli)
Supplier: BioVision
4001-20 4001-1000 4001-100
10005-208EA 607.09 USD
10005-208 10005-212 10005-210
Human Recombinant BCA-1 (from E. coli)
A cytokine selectively chemotactic for B cells
Determined by its ability to chemoattract human B cells using a concentration range of 1.0-10 ng/ml.
Human BCA-1 (B Cell-Attracting chemokine 1), also known as BLC or CXCL13, is a recently identified new CXC chemokines. Human BCA-1 is a highly effective attractant for human blood B lymphocytes, but was inactive on freshly isolated or IL-2 stimulated T lymphocytes, monocytes and neutrophils. The human BCA-1 is a 10.1 kDa protein containing 85 amino acid residues.