Would you like to log in again?
TNF-alpha, rat recombinant, BioVision
Supplier: BioVision
1052-10 1052-1000 1052-50
10003-018EA 147.52 USD InStock
10003-018 10003-020 10003-022
TNF-alpha, rat recombinant, BioVision
Proteins and Peptides
A potent cytokine exerting cytotoxic effects on a wide range of cells
The ED₅₀ as determined by the cytolysis of murine L929 cells in the presence of actinomycin D is < 0.05 ng/ml, corresponding to a specific activity of > 2 x 10⁷ units/mg.
Tumor Necrosis Factor-α (TNF-α) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Rat TNF-α is a 17 kDa protein containing 157 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty