TNF-alpha, murine recombinant, BioVision
1051-50 1051-1000 1051-10
10003-012EA 157.61 USD
10003-012 10003-014 10003-016
TNF-alpha, murine recombinant, BioVision
A potent cytokine exerting cytotoxic effects on a wide range of cells
The ED₅₀ as determined by the cytolysis of murine L929 cells in the presence of actinomycin D is < 0.1 ng/ml, corresponding to a specific activity of > 1 x 10⁷ units/mg.
Tumor Necrosis Factor-α TNF-α) is a potent lymphoid factor that exerts cytotoxic effects on a wide range of tumor cells and certain other target cells. Murine TNF-α is a 17.5 kDa protein containing 156 amino acid residues.