Would you like to log in again?
Procathepsin K, rat recombinant, BioVision
Supplier: BioVision
1029-50 1029-10 1029-1000
10003-004EA 921.85 USD InStock
10003-004 10003-000 10003-002
Procathepsin K, rat recombinant, BioVision
Proteins and Peptides
Predominant proteinase responsible for the resorption of the bone matrix.
The specific activity is >400 mU/mg (1U = 1 µmole/min/mg) as measured by Cathepsin K Activity Assay Kit (Catalog # K141-100).
Cathepsin K, a member of the papain cysteine proteinase family, is the predominant proteinase responsible for the resorption of the bone matrix. Cathepsin cleaves proteins such as collagen type I, collagen type II and osteonectin, thereby playing a role in bone remodeling and resorption in osteoporosis, osteolytic bone metastasis and rheumatoid arthritis (Bromme and Okamoto, 1995; Drake, F. et al 1996; Bossard et al, 1996). Cathepsin K is produced as an inactive proenzyme (35.4 kDa) that is converted to its mature active form (23.5 kDa) by proteolytic cleavage of its 99-amino-acid propeptide domain. The in-vitro processing of procathepsin K to mature cathepsin K is autocatalytic.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty