Would you like to log in again?
Procathepsin K, mouse recombinant, BioVision
Supplier: BioVision
1027-1000 1027-10 1027-50
10002-992EA 8188.5 USD InStock
10002-992 10002-990 10002-994
Procathepsin K, mouse recombinant, BioVision
Proteins and Peptides
Predominant proteinase responsible for the resorption of the bone matrix.
Cathepsin K is a member of the papain cysteine proteinase family and has been identified as the predominant proteinase responsible for the resorption of the bone matrix. The enzyme cleaves proteins such as collagen type I, collagen type II and osteonectin and therefore plays a role in bone remodeling and resorption in diseases such as osteoporosis, osteolytic bone metastasis and rheumatoid arthritis (Bromme and Okamoto, 1995; Drake, F. et al 1996; Bossard et al, 1996). Cathepsin K is synthesized as an inactive proenzyme (35.1 kDa) that is converted to its mature active form (23.6 kDa) by proteolytic cleavage of its 99-amino-acid propeptide domain. The in-vitro processing of procathepsin K to mature cathepsin K is autocatalytic.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty