Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Conductive Hanging Rail and Louvered Panel Systems, Quantum Storage Systems

Supplier: QUANTUM STORAGE SYSTEMS
QBR-1819CO QRU-12SCO QBR-3612CO QLP-4861CO QRU-16SCO QBR-3619CO QSS-3666HCO QLP-3612CO QLP-3619CO QSS-1866HCO QLP-1861CO QBR-2721CO QMD-36HCO QRU-12DCO QLP-4819CO QLP-2721CO QRU-16DCO QDS-3666HCO QLP-1819CO QLP-3661CO
Accepts all sizes of Conductive Hang and Stack Bins.

Ensure all racks and panels are grounded properly and suited to your specific application.



 

ORDER

Expand All / Collapse All
Description Dimensions Supplier No. VWR Catalog Number Unit Price Quantity
Conductive Hanging Bin System; Single Sided 12 Rail Systems 36 x 15 x 53" QRU-12SCO 10804-570 Each Retrieving
Conductive Hanging Bin System; Single Sided 16 Rail Systems 36 x 15 x 53" QRU-16SCO 10804-430 Each Retrieving
Conductive Hanging Bin System; Double Sided 12 Rail System 36 x 20 x 53" QRU-12DCO 10804-432 Each Retrieving
Conductive Hanging Bin System; Double Sided 16 Rail System 36 x 20 x 53" QRU-16DCO 10804-434 Each Retrieving
Conductive Hanging Bin System; Conductive Bench Rack 18 x 8 x 19" QBR-1819CO 10804-270 Each Retrieving
Conductive Hanging Bin System; Conductive Bench Rack 27 x 8 x 21" QBR-2721CO 10804-272 Each Retrieving
Conductive Hanging Bin System; Conductive Bench Rack 36 x 8 x 12" QBR-3612CO 10804-438 Each Retrieving
Conductive Hanging Bin System; Conductive Bench Rack 36 x 8 x 19" QBR-3619CO 10804-440 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 18 x 19" QLP-1819CO 10804-258 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 18 x 61" QLP-1861CO 10804-260 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 27 x 21" QLP-2721CO 10804-262 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 36 x 12" QLP-3612CO 10804-264 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 36 x 19" QLP-3619CO 10804-266 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 36 x 61" QLP-3661CO 10805-428 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 48 x 19" QLP-4819CO 10804-268 Each Retrieving
Conductive Hanging Bin System; Conductive Louvered Panel 48 x 61" QLP-4861CO 10805-430 Each Retrieving
Conductive Hanging Bin System; Single Sided Louvered Panel Rack 18 x 12 1/2 x 66" QSS-1866HCO 10805-432 Each Retrieving
Conductive Hanging Bin System; Single Sided Louvered Panel Rack 36 x 12 1/2 x 66" QSS-3666HCO 10805-434 Each Retrieving
Conductive Hanging Bin System; Double Sided Louvered Panel Rack 36 x 25 x 66" QDS-3666HCO 10805-436 Each Retrieving
Conductive Hanging Bin System; Double Sided Mobile Louvered Panel Rack 36 x 25 x 72" QMD-36HCO 10805-438 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

ACCESSORIES

Expand All / Collapse All

Accessories
Description Supplier No. VWR Catalog Number Unit Price Quantity
Mobile Kit for Double Sided Rail System QRU-MOBCO 10804-436 Each Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty