VWR® Performance Glass and Solid Door Stainless Steel Laboratory Refrigerators with Natural Refrigerants
HCPP-72G-TS
10819-448EA 21599.72 USD
10819-448
VWR® Performance Glass and Solid Door Stainless Steel Laboratory Refrigerators with Natural Refrigerants
Refrigerators
The VWR® Glass and Solid Door Stainless Steel Laboratory Refrigerators are purpose built for scientific and medical applications.
  • Stainless steel construction
  • HFC-free refrigerants
  • Remote alarm functions
  • Precise microprocessor temperature controller
Units feature a digital microprocessor temperature controller allowing for the precise temperature management necessary for critical samples and supplies
HFC-free refrigerants (or refrigerants not containing hydrofluorocarbons) vastly reduce the Global Warming Potential (GWP) and eliminate any potential for ozone depletion previously caused by the use of HFC-based refrigerants.
Remote alarm contacts enable end users to tie these units into existing alarm systems to notify appropriate personnel of alarm events as soon as possible. Forced-draft air circulation keeps cold air moving throughout the cabinet to ensure superior temperature uniformity and recovery after door openings. The cycle-defrost design allows for the self-maintenance necessary in all refrigerators to keep evaporator coils from freezing; but without the product-damaging spikes in temperature stemming from auto-defrost cycles. Height does not include 12.7 cm (5") for casters.
A forced air directional refrigeration system provides superior temperature uniformity and recovery after door openings. An automatic cycle-defrost design prevents product damaging temperature spikes. Multi-Point temperature mapping via NIST traceable data acquisition, 15 probes in air and 3 probes in sample solution for 360 minutes with temperature readings every 60 seconds
Certifications: UL/C-UL listed.
Ordering information: Units are supplied with a three-year parts and labor warranty, with an additional two years on compressor parts.