Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EcoClean® 35 Cotton Nonwoven Cleanroom Wiper, Berkshire

Supplier: BERKSHIRE CORP
ECW35.050530
EcoClean® is 100% biodegradable and renewable. It is made from 100% natural fiber, that contains no binders or additives, and it is manufactured by a proprietary process for low fiber release and high strength. EcoClean® 35 can be used in critical pharmaceutical, medical device, and semiconductor applications, where absorbency and softness are required.

Lightweight, high absorbency design reduces landfill waste impact when compared to more traditional heavier weight products

Applications include cleaning high temperature production equipment and diffusion furnaces, critical industrial applications, where absorbency and softness are critical, as well as optical materials.

Cleaning of less critical areas and manufacturing processes in regulated environments. Apertured surface aids in particle pick up and entrapment.

Certifications: Certified for contact with food per EC Regulation 1935(2004) - FDA 21 CFR - Food contact compliant.

Ordering information: 100 sheets per pack, 30 packs per case (double packed).




 

ORDER

Expand All / Collapse All
Size Packaging Supplier No. VWR Catalog Number Unit Price Quantity
12.5 x 12.5 cm (5 x 5") Quarter folded ECW35.050530 89097-478 Case of 3000 Retrieving
Pack of 100 Retrieving
Small Business Enterprise
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty