Would you like to log in again?
Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Supplier: BERKSHIRE CORP
CH809.10
89089-180CS 598.93 USD InStock
89089-180
Choice® Cleanroom Laundered 100% Polyester Knit Wipe, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
Knife cut edge for reduced scratching potential on sensitive surfaces
Chemically compatible with IPA, acetone and other solvents.
Applications include chamber cleaning and CMP processing, stencil and print roll cleaning applications, cleaning of medical device products, applying and removing cleaning and disinfecting solutions, CD and DVD manufacturing processes. Steam autoclavable for aseptic environments.
Certifications: Complies with RoHS and REACH requirements.
Ordering information: 150 sheets per pack, 10 packs per case. (double packed). Laundered and packaged in ISO class 4 (class 10) cleanroom.
Order Now
ORDER
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty