Would you like to log in again?
IL-4, rat recombinant, BioVision
Supplier: BioVision
4139-50 4139-1000 4139-10
10006-150EA 643.75 USD InStock
10006-150 10006-148 10006-146
IL-4, rat recombinant, BioVision
Proteins and Peptides
A key regulator in humoral and adaptive immunity
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of con A-activated rat spleen cells is < 1.5 ng/ml, corresponding to a specific activity of > 6.6 x 10⁵ units/mg.
IL4 is a pleiotropic cytokine and a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B cells and T cells. It is produced by activated T cells and is a ligand for interleukin 4 receptor. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal
transducer and activator of transcription, has been shown to play a central role in mediating the immune regulatory signal of this cytokine. Two alternatively spliced transcript variants of this gene encoding distinct isoforms have been reported.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty