Would you like to log in again?
IL-2, rat recombinant, BioVision
Supplier: BioVision
4133-1000 4133-100
10006-136EA 2832.5 USD InStock
10006-136 10006-134
IL-2, rat recombinant, BioVision
Proteins and Peptides
A regulator of lymphocyte activities
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells is <0.4 ng/ml, corresponding to a specific activity of >2.5 x 10⁶ units/mg.
Interleukin-2 (IL-2) is a potent lymphoid cell growth factor that exerts its biological activity primarily on T cells. The recombinant rat IL-2 is a 15.3 kDa protein containing 134 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty