Would you like to log in again?
IL-2, murine recombinant, BioVision
Supplier: BioVision
4132-1000 4132-100 4132-20
10006-130EA 3769.8 USD InStock
10006-130 10006-128 10006-132
IL-2, murine recombinant, BioVision
Proteins and Peptides
A regulator of lymphocyte activities
The ED₅₀ range is 0.1-0.4 ng/ml, as determined by the dose-dependent stimulation of the proliferation of murine CTLL-2 cells.
Interleukin-2 (IL-2) is a potent lymphoid cell growth factor that exerts its biological activity primarily on T cells. The recombinant Murine IL-2 is a 17.2 kDa protein containing 149 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty