Would you like to log in again?
IL-1 beta, murine recombinant, BioVision
Supplier: BioVision
4129-50 4129-10 4129-1000
10006-114EA 612.85 USD InStock
10006-114 10006-110 10006-112
IL-1 beta, murine recombinant, BioVision
Proteins and Peptides
An important mediator of inflammatory responses
The ED₅₀ determined by the dose-dependent stimulation of murine D10S cells is <0.002 ng/ml, corresponding to a specific activity of >5 x 10⁸ units/mg.
IL-1β is a proinflammatory cytokine produced in a variety of cells including monocytes, tissue macrophages, keratinocytes and other epithelial cells. Both IL-1α and IL-1β binds to the same receptor and has similar if not identical biological properties. These cytokines have a broad range of activities including, stimulation of thymocyte proliferation, by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity and the ability to stimulate the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant murine IL-1β is a 17.5 kDa protein containing 153 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty