Would you like to log in again?
IL-1 beta, human recombinant, BioVision
Supplier: BioVision
4128-1000 4128-10 4128-50
10005-634 10005-632 10006-108
IL-1 beta, human recombinant, BioVision
An important mediator of inflammatory responses
The ED₅₀ as determined by the dose-dependent stimulation of thymidine uptake by murine C3H/HeJ thymocytes is <0.1 ng/ml, corresponding to a specific activity of >1 x 10⁷ units/mg.
IL-1β is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses including the activation of B and T cells. Recombinant human IL-1β is a 17.0 kDa protein containing 153 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty