Would you like to log in again?
IL-1 alpha,rat recombinant, BioVision
Supplier: BioVision
4127-10 4127-1000
10005-628 10005-630
IL-1 alpha,rat recombinant, BioVision
Activates TNF-α and plays a critical role in immune response regulation
The ED₅₀ as determined by the dose-dependent stimulation of the proliferation of murine D10S cells is <0.005 ng/ml, corresponding to a specific activity of >2 x 10⁹ units/mg.
Interleukin-1 α (IL-1α is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses. Rat IL-1α is a 17.7 kDa protein containing 155 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty