Human Recombinant IL1 alpha (from E. coli)
Supplier: BioVision
4125-50 4125-10 4125-1000
10006-096EA 210.14 USD
10006-096 10006-100 10006-098
Human Recombinant IL1 alpha (from E. coli)
Activates TNF-α and plays a critical role in immune response regulation
The ED₅₀ as determined by the dose-dependent stimulation of murine D10S cells is <0.001 ng/ml, corresponding to a specific activity of >1 x 10⁹ units/mg.
IL-1α is a potent immuno-modulator that mediates a wide range of immune and inflammatory responses. BioVision’s Human IL-1α is an 18.0 kDa protein containing 159 amino acid residues.