Would you like to log in again?
IFN-gamma, rat recombinant, BioVision
Supplier: BioVision
4118-1000 4118-20 4118-100
10006-078EA 1854 USD InStock
10006-078 10006-080 10006-076
IFN-gamma, rat recombinant, BioVision
Proteins and Peptides
A critical cytokine for innate and adaptive immunity
The ED₅₀ as determined by a cytopathic affect inhibition assay with murine L929 cells challenged with EMC virus was < 0.1 ng/ml, corresponding to a specific activity of > 1 x 10⁷ units/mg.
IFN-γ is a lymphoid factor that possesses potent anti-viral activity. It has also been shown to stimulate macrophages and NK cells. Rat IFN-g is a 15.6 kDa protein containing 135 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty