Would you like to log in again?
IFN-gamma, murine recombinant, BioVision
Supplier: BioVision
4117-1000 4117-100 4117-20
10006-072 10006-070 10006-074
IFN-gamma, murine recombinant, BioVision
A critical cytokine for innate and adaptive immunity
The ED₅₀ as determined by a cytopathic affect inhibition assay with murine NIH 3T3 cells challenged with VSV virus is < 0.1 ng/ml, corresponding to a specific activity of > 1 x 10⁷ units/mg.
IFN-γ is an acid-labile interferon produced by CD4 and CD8 T lymphocytes as well as activated NK cells. IFN-γ receptors are present in most immune cells, which respond to IFN-γ signaling by increasing the surface expression of class I MHC proteins. This promotes the presentation of antigen to T-helper (CD4+) cells. IFN-γ signaling in antigen-presenting cells and antigen-recognizing B and T lymphocytes regulate the antigen-specific phases of the immune response. Additionally, IFN-γ stimulates a number of lymphoid cell functions including the anti-microbial and anti-tumor responses of macrophages, NK cells, and neutrophils. Recombinant murine IFN-γ is a 15.6 kDa protein containing 134 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty