Would you like to log in again?
I-309, human recombinant, BioVision
Supplier: BioVision
4113-1000 4113-10
10006-062 10006-060
I-309, human recombinant, BioVision
A monocyte chemoattractant
The biological activity of the recombinant human I-309 was determined by a chemotaxis assay using total human T cell population. Significant chemotaxis was achieved at a concentration range of 1-100 ng/ml.
Human I-309 is a small glycoprotein secreted by activated T lymphocytes. It is structurally related to other CC chemokines. The recombinant human I-309 is an 8.5 kDa protein containing 74 amino acid residues.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty