Would you like to log in again?
Recombinant Mouse M-CSF (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-8983-U010 21-8983-U002
10821-234EA 106.74 USD InStock
10821-234 10821-232
Recombinant Mouse M-CSF (Carrier-free), Tonbo Biosciences
Proteins and Peptides
M-CSF is a hematopoietic growth factor that drives macrophage, monocyte, and bone marrow progenitor cell proliferation, differentiation, and survival. M-CSF increases macrophage and monocyte phagocytosis, chemotaxis, and tumoricidal activity making it an important therapeutic agent. M-CSF is used clinically for treatment of cancer, infectious diseases, and atherosclerosis. It is also used in recovery following bone marrow transplants. M-CSF is elaborated by multiple cells including lymphocytes, monocytes, fibroblasts, endothelial cells, myoblasts, and osteoblasts. M-CSF has also been associated with dermal thickening, pregnancy maintenance and female and male fertility.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty