Would you like to log in again?
Recombinant Human sIL-2Rα (sCD25) (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-8702-U025 21-8702-U100 21-8702-U500 21-8702-U005
10830-248 10830-250 10830-252 10830-246
Recombinant Human sIL-2Rα (sCD25) (Carrier-free), Tonbo Biosciences
The high-affinity IL-2 receptor is a trimeric complex made up of IL-2R alpha (CD25), IL-2R beta (CD122) and the common gamma chain (IL-2R gamma, CD132). The alpha chain is cleaved from the cell membrane through proteolytic processing to release soluble IL-2R alpha. IL-2R alpha binds IL-2 with low affinity and is not essential for IL-2 signaling, but when combined with the beta and gamma chains it greatly increases affinity for IL-2. IL-2R alpha is expressed during early T and B lymphocyte development and is also induced during mature T and B cell activation. It has been reported to enhance Th17 responses in vitro.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty