Would you like to log in again?
Recombinant Mouse GDNF (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-8505-U010 21-8505-U500 21-8505-U002 21-8505-U100
10830-240EA 205.62 USD InStock
10830-240 10830-244 10830-238 10830-242
Recombinant Mouse GDNF (Carrier-free), Tonbo Biosciences
Proteins and Peptides
Glial Cell derived Neurotrophic Factor (GDNF) is a highly conserved neurotrophic factor. It is a novel member of the TGF-beta superfamily that has been shown to promote survival of various neuronal subpopulations in both the central as well as the peripheral nervous systems at different stages of development. Upregulated expression of GNDF was observed in various cancer cell lines and correlates with malignant cancer progression, suggesting a role of GDNF in carcinogenesis. The mature form of GDNF is a secreted protein that exists as a disulfide-linked homodimeric glycoprotein.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty