Would you like to log in again?
Recombinant Human IL-36β (IL-1F8) (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-8372-U100 21-8372-U010 21-8372-U500
10830-234EA 1137.19 USD InStock
10830-234 10830-232 10830-236
Recombinant Human IL-36β (IL-1F8) (Carrier-free), Tonbo Biosciences
Proteins and Peptides
IL-36 beta, also known as IL-1F8, is a member of the IL-1 family of cytokines and binds to the IL-1R6 (IL-1Rrp2) receptor. It is reported to be expressed by resting and activated monocytes and B cells and at low levels in various tissues. It is highly expressed in psoriatic plaques. IL-36 beta displays proinflammatory effects and, like other IL-36 cytokines, can activate NF-kB and MAPK signaling pathways.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty