Would you like to log in again?
Recombinant Human sTRAIL Receptor 2 (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7160-U500 21-7160-U050 21-7160-U010 21-7160-U100
10821-210EA 983.02 USD InStock
10821-210 10821-206 10821-204 10821-208
Recombinant Human sTRAIL Receptor 2 (Carrier-free), Tonbo Biosciences
Proteins and Peptides
TRAIL Receptor 2 (DR5, TNFRSF10B, CD262, TRICK2) is a type I transmembrane protein belonging to the TNF receptor superfamily. It contains a cytoplasmic death domain and is one of the four receptors that bind TRAIL to initiate apoptosis. TRAIL R2 mediated apoptosis involves a signaling complex that includes FADD and caspases 8 and 10. TRAIL R2 expression can be induced by NF-kappaB.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty