Would you like to log in again?
Recombinant Human sTRAIL Receptor 1 (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7159-U100 21-7159-U050 21-7159-U010 21-7159-U500
10821-200EA 318.81 USD InStock
10821-200 10821-198 10821-196 10821-202
Recombinant Human sTRAIL Receptor 1 (Carrier-free), Tonbo Biosciences
Proteins and Peptides
TRAIL Receptor 1 (DR4, TNFRSF10A, CD261, APO2) is a type I transmembrane protein belonging to the TNF receptor superfamily. It contains a cytoplasmic death domain and is one of the four receptors that bind TRAIL to initiate apoptosis. TRAIL R1 mediated apoptosis involves a signaling complex that includes FADD and caspases 8 and 10. TRAIL R1 expression can be induced by NF-kappaB.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty