Would you like to log in again?
Recombinant Human LIGHT (Carrier-free), Tonbo Biosciences
Supplier: Tonbo Biosciences
21-7155-U003 21-7155-U015
10821-168EA 86.98 USD InStock
10821-168 10821-170
Recombinant Human LIGHT (Carrier-free), Tonbo Biosciences
Proteins and Peptides
LIGHT, also known as TNFSF14, CD258 and HVEM-L, is a type II transmembrane protein belonging to the TNF family of ligands. It is produced by activated T-cells and signals via the lymphotoxin beta receptor (LTbetaR) and the herpes virus entry mediator (HVEM). It is a potent initiator of T cell co-stimulation signals that play a role in tumor and allograft rejection. LIGHT is reported to work together with IFN-g to activate caspase activity, resulting in cleavage of Bcl-2.
Order Now
ORDER
SPECIFICATIONS
RELATED INFORMATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty